siRNA Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053662.3 | NP_446114.1 | 801 | |||
| XM_008761029.1 | XP_008759251.1 | 1877 | ||||
| GenBank mRNA | AF030091.3 | - | 801 | |||
| BC085686.1 | - | 855 | ||||
| BC128699.1 | - | 895 | ||||
| BC161817.1 | - | 747 | 9022416 | |||
| FQ213168.1 | - | 3 | 225 | |||
| FQ232436.1 | - | 838 | ||||

Ccnl1
Genomic Map