siRNA Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053665.1 | NP_446117.1 | 1 | 370 | ||
| GenBank mRNA | AF068202.1 | - | 1 | 370 | ||
| AF092523.1 | - | 1 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap1| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053665.1 | NP_446117.1 | 1 | 370 | ||
| GenBank mRNA | AF068202.1 | - | 1 | 370 | ||
| AF092523.1 | - | 1 | 370 | |||