siRNA Details
| Gene Symbol: CD160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007053.3 | NP_008984.1 | 4 | 405 | ||
| XM_005272929.3 | XP_005272986.1 | 3 | 433 | |||
| XM_011509104.2 | XP_011507406.1 | 4 | 539 | |||
| GenBank mRNA | AF060981.1 | - | 4 | 328 | ||
| BC014465.1 | - | 4 | 331 | 4849145 | ||
| CR541867.1 | - | 2 | 113 | |||
| CR541888.1 | - | 2 | 113 | |||
| EU016100.1 | - | 2 | 113 | |||

CD160
Genomic Map