siRNA Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053682.2 | NP_446134.2 | 12 | 1414 | ||
| XM_006254339.2 | XP_006254401.1 | 12 | 1435 | |||
| GenBank mRNA | AF151784.1 | - | 12 | 1278 | ||
| BC081751.1 | - | 12 | 1414 | 7108995 | ||
| Gene Symbol: LOC100910779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751732.3 | XP_003751780.1 | 1652 | |||
| XM_006254369.2 | XP_006254431.1 | 1435 | ||||

Yme1l1
Genomic Map