siRNA Details
| Gene Symbol: Dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053706.1 | NP_446158.1 | 2 | 536 | ||
| XM_008760301.1 | XP_008758523.1 | 2 | 626 | |||
| GenBank mRNA | AF379608.1 | - | 2 | 536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrt1| Gene Symbol: Dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053706.1 | NP_446158.1 | 2 | 536 | ||
| XM_008760301.1 | XP_008758523.1 | 2 | 626 | |||
| GenBank mRNA | AF379608.1 | - | 2 | 536 | ||