siRNA Details
| Gene Symbol: Slc5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053715.2 | NP_446167.2 | 1 | 763 | ||
| GenBank mRNA | AJ001290.1 | - | 1 | 763 | ||
| AY231162.1 | - | 1 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a3| Gene Symbol: Slc5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053715.2 | NP_446167.2 | 1 | 763 | ||
| GenBank mRNA | AJ001290.1 | - | 1 | 763 | ||
| AY231162.1 | - | 1 | 704 | |||