siRNA Details
| Gene Symbol: Mllt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053718.2 | NP_446170.1 | 2 | 507 | ||
| GenBank mRNA | AJ006295.1 | - | 2 | 507 | ||
| BC129089.1 | - | 211 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mllt3| Gene Symbol: Mllt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053718.2 | NP_446170.1 | 2 | 507 | ||
| GenBank mRNA | AJ006295.1 | - | 2 | 507 | ||
| BC129089.1 | - | 211 | ||||