siRNA Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053748.1 | NP_446200.1 | 3 | 388 | ||
| XM_006230633.2 | XP_006230695.1 | 4 | 575 | |||
| GenBank mRNA | BC107673.1 | - | 4 | 399 | 7938129 | |
| D89340.2 | - | 3 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp3| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053748.1 | NP_446200.1 | 3 | 388 | ||
| XM_006230633.2 | XP_006230695.1 | 4 | 575 | |||
| GenBank mRNA | BC107673.1 | - | 4 | 399 | 7938129 | |
| D89340.2 | - | 3 | 388 | |||