siRNA Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053748.1 | NP_446200.1 | 16 | 2020 | ||
| XM_006230633.2 | XP_006230695.1 | 17 | 2207 | |||
| GenBank mRNA | BC107673.1 | - | 17 | 2031 | 7938129 | |
| D89340.2 | - | 16 | 2020 | |||

Dpp3
Genomic Map