siRNA Details
| Gene Symbol: Pkia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053772.2 | NP_446224.1 | 5 | 1456 | ||
| XM_006232153.2 | XP_006232215.1 | 3 | 1652 | |||
| XM_006232155.2 | XP_006232217.1 | 4 | 1303 | |||
| XM_006232156.2 | XP_006232218.1 | 4 | 3355 | |||
| XM_008760857.1 | XP_008759079.1 | 4 | 1780 | |||
| GenBank mRNA | FQ211701.1 | - | 4 | 1363 | ||
| L02615.1 | - | 3 | 1137 | |||

Pkia
Genomic Map