siRNA Details
| Gene Symbol: Serpini1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053779.1 | NP_446231.1 | 3 | 467 | ||
| XM_008761076.1 | XP_008759298.1 | 4 | 754 | |||
| GenBank mRNA | AF193014.1 | - | 3 | 467 | ||
| AF193015.1 | - | 3 | 473 | |||
| BC061536.1 | - | 3 | 481 | 6920827 | ||

Serpini1
Genomic Map