siRNA Details
| Gene Symbol: Sec14l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053801.2 | NP_446253.2 | 5 | 421 | ||
| XM_006251154.2 | XP_006251216.1 | 5 | 410 | |||
| GenBank mRNA | AF309558.1 | - | 5 | 378 | ||
| BC089785.1 | - | 5 | 421 | 7322689 | ||

Sec14l2
Genomic Map