siRNA Details
| Gene Symbol: Snap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053810.3 | NP_446262.3 | 2, 3 | 516 | ||
| GenBank mRNA | AF035822.1 | - | 2, 3 | 526 | ||
| AF260577.1 | - | 2, 3 | 439 | |||
| BC091693.1 | - | 2, 3 | 516 | 7134635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snap29| Gene Symbol: Snap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053810.3 | NP_446262.3 | 2, 3 | 516 | ||
| GenBank mRNA | AF035822.1 | - | 2, 3 | 526 | ||
| AF260577.1 | - | 2, 3 | 439 | |||
| BC091693.1 | - | 2, 3 | 516 | 7134635 | ||