siRNA Details
| Gene Symbol: Nrxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053817.2 | NP_446269.2 | 2 | 1090 | ||
| XM_008764731.1 | XP_008762953.1 | 1 | 343 | |||
| GenBank mRNA | L14851.1 | - | 2 | 1090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrxn3| Gene Symbol: Nrxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053817.2 | NP_446269.2 | 2 | 1090 | ||
| XM_008764731.1 | XP_008762953.1 | 1 | 343 | |||
| GenBank mRNA | L14851.1 | - | 2 | 1090 | ||