siRNA Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053881.1 | NP_446333.1 | 5 | 630 | ||
| XM_006245134.2 | XP_006245196.1 | 5 | 698 | |||
| GenBank mRNA | FQ211652.1 | - | 5 | 689 | ||
| U40652.1 | - | 5 | 596 | |||
| X92563.1 | - | 5 | 630 | |||

Ptprn
Genomic Map