siRNA Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053948.4 | NP_446400.1 | 8 | 825 | ||
| XM_006230973.1 | XP_006231035.1 | 10 | 1110 | |||
| GenBank mRNA | BC060550.1 | - | 8 | 781 | 6890594 | |
| BC060595.1 | - | 10 | 1119 | 6888231 | ||
| FQ223906.1 | - | 10 | 1115 | |||
| FQ224086.1 | - | 8 | 858 | |||
| FQ230390.1 | - | 8 | 788 | |||
| FQ232354.1 | - | 8 | 829 | |||
| Z71925.1 | - | 8 | 826 | |||

Polr2g
Genomic Map