siRNA Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053949.1 | NP_446401.1 | 7 | 1838 | ||
| XM_006235867.2 | XP_006235929.1 | 7 | 1971 | |||
| XM_008762630.1 | XP_008760852.1 | 3 | 1159 | |||
| GenBank mRNA | AY669863.1 | - | 3 | 812 | ||
| Z96106.1 | - | 7 | 1838 | |||

Kcnh2
Genomic Map