siRNA Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053953.1 | NP_446405.1 | 8 | 1118 | ||
| GenBank mRNA | Z22812.1 | - | 8 | 1118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1r2| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053953.1 | NP_446405.1 | 8 | 1118 | ||
| GenBank mRNA | Z22812.1 | - | 8 | 1118 | ||