siRNA Details
| Gene Symbol: Spam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053967.2 | NP_446419.1 | 2 | 589 | ||
| GenBank mRNA | BC081748.1 | - | 2 | 530 | 7134081 | |
| X89999.1 | - | 2 | 593 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spam1| Gene Symbol: Spam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053967.2 | NP_446419.1 | 2 | 589 | ||
| GenBank mRNA | BC081748.1 | - | 2 | 530 | 7134081 | |
| X89999.1 | - | 2 | 593 | |||