siRNA Details
| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057118.2 | NP_476459.1 | 4 | 273 | ||
| XM_006242177.2 | XP_006242239.1 | 4 | 288 | |||
| XM_008765659.1 | XP_008763881.1 | 3 | 214 | |||
| GenBank mRNA | D38492.1 | - | 4 | 277 | ||

Cntn1
Genomic Map