siRNA Details
| Gene Symbol: Sept2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057148.2 | NP_476489.1 | 7 | 556 | ||
| XM_006245511.2 | XP_006245573.1 | 7 | 836 | |||
| XM_006245512.2 | XP_006245574.1 | 7 | 839 | |||
| XM_006245513.2 | XP_006245575.1 | 7 | 580 | |||
| XM_006245514.1 | XP_006245576.1 | 7 | 614 | |||
| GenBank mRNA | AB027561.1 | - | 6 | 517 | ||
| BC081745.1 | - | 7 | 556 | 7121592 | ||
| FQ212097.1 | - | 7 | 569 | |||

Sept2
Genomic Map