siRNA Details
| Gene Symbol: Xpnpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057155.3 | NP_476496.1 | 7 | 583 | ||
| XM_008773513.1 | XP_008771735.1 | 8 | 1023 | |||
| XM_008773514.1 | XP_008771736.1 | 7 | 886 | |||
| GenBank mRNA | AF359355.1 | - | 7 | 583 | ||
| BC074017.1 | - | 7 | 847 | 7098467 | ||

Xpnpep2
Genomic Map