siRNA Details
| Gene Symbol: St8sia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057156.1 | NP_476497.1 | 3 | 204 | ||
| GenBank mRNA | L13445.1 | - | 3 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia2| Gene Symbol: St8sia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057156.1 | NP_476497.1 | 3 | 204 | ||
| GenBank mRNA | L13445.1 | - | 3 | 204 | ||