siRNA Details
| Gene Symbol: Rps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078617.3 | NP_511172.1 | 2 | 191 | ||
| GenBank mRNA | BC058134.1 | - | 2 | 195 | 6918484 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps23| Gene Symbol: Rps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078617.3 | NP_511172.1 | 2 | 191 | ||
| GenBank mRNA | BC058134.1 | - | 2 | 195 | 6918484 | |