siRNA Details
| Gene Symbol: Sytl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080410.1 | NP_536335.1 | 15 | 1748 | ||
| XM_006257213.2 | XP_006257275.1 | 16 | 1956 | |||
| GenBank mRNA | AF419341.1 | - | 15 | 1748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytl4| Gene Symbol: Sytl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080410.1 | NP_536335.1 | 15 | 1748 | ||
| XM_006257213.2 | XP_006257275.1 | 16 | 1956 | |||
| GenBank mRNA | AF419341.1 | - | 15 | 1748 | ||