siRNA Details
| Gene Symbol: Bnip3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080888.1 | NP_543164.1 | 6 | 944 | ||
| XM_006252091.2 | XP_006252153.1 | 6 | 2745 | |||
| XM_008770733.1 | XP_008768955.1 | 6 | 2874 | |||
| GenBank mRNA | AB077364.1 | - | 6 | 944 | ||
| AF441118.1 | - | 6 | 1012 | |||
| BC081740.1 | - | 6 | 1098 | |||
| BC098658.1 | - | 6 | 978 | 7324593 | ||
| FQ230363.1 | - | 6 | 937 | |||
| FQ235252.1 | - | 6 | 963 | |||

Bnip3l
Genomic Map