siRNA Details
| Gene Symbol: Rcvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080901.1 | NP_543177.1 | 3 | 830 | ||
| GenBank mRNA | AY063482.1 | - | 3 | 830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcvrn| Gene Symbol: Rcvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080901.1 | NP_543177.1 | 3 | 830 | ||
| GenBank mRNA | AY063482.1 | - | 3 | 830 | ||