siRNA Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080908.1 | NP_543184.1 | 2 | 172 | ||
| XM_008771422.1 | XP_008769644.1 | 2 | 257 | |||
| GenBank mRNA | AY059414.1 | - | 2 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc28a3| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080908.1 | NP_543184.1 | 2 | 172 | ||
| XM_008771422.1 | XP_008769644.1 | 2 | 257 | |||
| GenBank mRNA | AY059414.1 | - | 2 | 172 | ||