siRNA Details
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080908.1 | NP_543184.1 | 2 | 175 | ||
XM_008771422.1 | XP_008769644.1 | 2 | 260 | |||
GenBank mRNA | AY059414.1 | - | 2 | 175 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080908.1 | NP_543184.1 | 2 | 175 | ||
XM_008771422.1 | XP_008769644.1 | 2 | 260 | |||
GenBank mRNA | AY059414.1 | - | 2 | 175 |