siRNA Details
| Gene Symbol: Rtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009953.2 | NP_001009953.2 | 7 | 810 | ||
| NM_080909.2 | NP_543185.2 | 8 | 2984 | |||
| XM_006230649.2 | XP_006230711.1 | 9 | 3056 | |||
| XM_006230650.2 | XP_006230712.1 | 7 | 780 | |||
| GenBank mRNA | AF442357.1 | - | 7 | 814 | ||
| AF442358.1 | - | 7 | 814 | |||
| AY164698.1 | - | 7 | 814 | |||
| AY496443.1 | - | 8 | 2984 | |||
| BC062068.1 | - | 7 | 808 | |||
| BC107448.1 | - | 7 | 810 | |||
| BC129077.1 | - | 7 | 818 | |||

Rtn3
Genomic Map