siRNA Details
| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080910.3 | NP_543186.1 | 3, 4 | 314 | ||
| XM_006250852.2 | XP_006250914.1 | 3, 4 | 362 | |||
| XM_006250853.2 | XP_006250915.1 | 3, 4 | 333 | |||
| XM_006250854.2 | XP_006250916.1 | 3, 4 | 363 | |||
| GenBank mRNA | BC072508.1 | - | 3, 4 | 304 | 7103198 | |
| BC085711.1 | - | 3, 4 | 314 | 7192781 | ||
| D37979.1 | - | 1, 2 | 219 | |||

Paics
Genomic Map