siRNA Details
| Gene Symbol: Skap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130413.1 | NP_569097.1 | 5 | 479 | ||
| XM_006236477.2 | XP_006236539.1 | 5 | 464 | |||
| XM_006236478.2 | XP_006236540.1 | 5 | 464 | |||
| GenBank mRNA | AF302132.1 | - | 5 | 479 | ||
| BC070949.1 | - | 5 | 426 | 7099436 | ||

Skap2
Genomic Map