siRNA Details
| Gene Symbol: Tnfrsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130426.4 | NP_569110.1 | 2 | 227 | ||
| GenBank mRNA | AF420214.1 | - | 2 | 98 | ||
| AF498039.2 | - | 2 | 227 | |||
| AY191268.1 | - | 227 | ||||
| AY191269.1 | - | 227 | ||||
| AY344841.1 | - | 2 | 140 | |||

Tnfrsf1b
Genomic Map