siRNA Details
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130433.1 | NP_569117.1 | 2 | 314 | ||
GenBank mRNA | FQ216726.1 | - | 2 | 177 | ||
FQ228855.1 | - | 2 | 179 | |||
X05341.1 | - | 2 | 314 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130433.1 | NP_569117.1 | 2 | 314 | ||
GenBank mRNA | FQ216726.1 | - | 2 | 177 | ||
FQ228855.1 | - | 2 | 179 | |||
X05341.1 | - | 2 | 314 |