siRNA Details
Gene Symbol: Fgf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130754.1 | NP_570110.1 | 2 | 277 | ||
GenBank mRNA | AB078777.1 | - | 2 | 277 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130754.1 | NP_570110.1 | 2 | 277 | ||
GenBank mRNA | AB078777.1 | - | 2 | 277 |