siRNA Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130755.1 | NP_570111.1 | 2, 3 | 171 | ||
| GenBank mRNA | AF461496.1 | - | 2, 3 | 171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cs| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130755.1 | NP_570111.1 | 2, 3 | 171 | ||
| GenBank mRNA | AF461496.1 | - | 2, 3 | 171 | ||