siRNA Details
| Gene Symbol: Samsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130821.1 | NP_570834.1 | 3 | 268 | ||
| GenBank mRNA | AB067679.1 | - | 3 | 268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samsn1| Gene Symbol: Samsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130821.1 | NP_570834.1 | 3 | 268 | ||
| GenBank mRNA | AB067679.1 | - | 3 | 268 | ||