siRNA Details
| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130828.2 | NP_570841.2 | 1 | 257 | ||
| GenBank mRNA | AB091379.1 | - | 1 | 196 | ||
| AY039651.1 | - | 1 | 257 | |||
| BC098649.1 | - | 1 | 239 | 7127895 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nphs2| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130828.2 | NP_570841.2 | 1 | 257 | ||
| GenBank mRNA | AB091379.1 | - | 1 | 196 | ||
| AY039651.1 | - | 1 | 257 | |||
| BC098649.1 | - | 1 | 239 | 7127895 | ||