siRNA Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131904.1 | NP_571979.1 | 4 | 387 | ||
| GenBank mRNA | AY039679.1 | - | 4 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgea5| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131904.1 | NP_571979.1 | 4 | 387 | ||
| GenBank mRNA | AY039679.1 | - | 4 | 387 | ||