siRNA Details
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131905.1 | NP_571980.1 | 2 | 451 | ||
GenBank mRNA | AF333099.1 | - | 2 | 451 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131905.1 | NP_571980.1 | 2 | 451 | ||
GenBank mRNA | AF333099.1 | - | 2 | 451 |