siRNA Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131913.1 | NP_571988.1 | 15, 16 | 1375 | ||
| GenBank mRNA | AF038591.1 | - | 15, 16 | 1375 | ||
| BC061758.1 | - | 16, 17 | 1485 | 5600093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpnpep1| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131913.1 | NP_571988.1 | 15, 16 | 1375 | ||
| GenBank mRNA | AF038591.1 | - | 15, 16 | 1375 | ||
| BC061758.1 | - | 16, 17 | 1485 | 5600093 | ||