siRNA Details
| Gene Symbol: Erg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133397.2 | NP_596888.1 | 9 | 1634 | ||
| GenBank mRNA | AB031088.1 | - | 9 | 1634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erg| Gene Symbol: Erg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133397.2 | NP_596888.1 | 9 | 1634 | ||
| GenBank mRNA | AB031088.1 | - | 9 | 1634 | ||