siRNA Details
| Gene Symbol: Ilk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133409.2 | NP_596900.1 | 4 | 451 | ||
| GenBank mRNA | AF329194.1 | - | 4 | 388 | ||
| BC062406.1 | - | 4 | 451 | 5623251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ilk| Gene Symbol: Ilk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133409.2 | NP_596900.1 | 4 | 451 | ||
| GenBank mRNA | AF329194.1 | - | 4 | 388 | ||
| BC062406.1 | - | 4 | 451 | 5623251 | ||