siRNA Details
Gene Symbol: Mmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133514.1 | NP_598198.1 | 1 | 126 | ||
GenBank mRNA | M65253.1 | - | 1 | 126 | ||
X05083.1 | - | 1 | 123 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133514.1 | NP_598198.1 | 1 | 126 | ||
GenBank mRNA | M65253.1 | - | 1 | 126 | ||
X05083.1 | - | 1 | 123 |