siRNA Details
| Gene Symbol: Svs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133516.2 | NP_598200.1 | 2 | 284 | ||
| GenBank mRNA | AF159097.1 | - | 1 | 53 | ||
| X01115.1 | - | 2 | 284 | |||
| Gene Symbol: LOC103694892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773873.1 | XP_008772095.1 | 2 | 319 | ||

Svs5
Genomic Map