siRNA Details
| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133518.1 | NP_598202.1 | 5 | 434 | ||
| XM_006249379.2 | XP_006249441.1 | 5 | 366 | |||
| XM_006249380.2 | XP_006249442.1 | 5 | 671 | |||
| XM_006249381.2 | XP_006249443.1 | 5 | 670 | |||
| GenBank mRNA | U12571.1 | - | 5 | 434 | ||

Rph3a
Genomic Map