siRNA Details
| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133537.2 | NP_598221.2 | 3 | 380 | ||
| GenBank mRNA | X13309.1 | - | 3 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc18| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133537.2 | NP_598221.2 | 3 | 380 | ||
| GenBank mRNA | X13309.1 | - | 3 | 369 | ||