siRNA Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133538.2 | NP_598222.2 | 8 | 910 | ||
| XM_006257427.2 | XP_006257489.1 | 7 | 778 | |||
| XM_006257428.2 | XP_006257490.1 | 7 | 896 | |||
| XM_006257430.2 | XP_006257492.1 | 6 | 709 | |||
| XM_006257431.2 | XP_006257493.1 | 4 | 417 | |||
| XM_008773388.1 | XP_008771610.1 | 7 | 1050 | |||
| XM_008773389.1 | XP_008771611.1 | 8 | 937 | |||
| XM_008773390.1 | XP_008771612.1 | 4 | 511 | |||
| XM_008773391.1 | XP_008771613.1 | 4 | 1118 | |||
| GenBank mRNA | AF448818.1 | - | 8 | 910 | ||

Il13ra2
Genomic Map