siRNA Details
Gene Symbol: ZNF195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130519.2 | NP_001123991.1 | 4 | 533 | ||
NM_001130520.2 | NP_001123992.1 | 4 | 533 | |||
NM_001242841.1 | NP_001229770.1 | 5 | 613 | |||
NR_046381.1 | 6 | 670 | ||||
XM_011520350.2 | XP_011518652.1 | 5 | 522 | |||
XM_011520351.2 | XP_011518653.1 | 6 | 893 | |||
XM_017018260.1 | XP_016873749.1 | 5 | 736 | |||
XM_017018261.1 | XP_016873750.1 | 5 | 618 | |||
XM_017018262.1 | XP_016873751.1 | 5 | 736 | |||
XM_017018263.1 | XP_016873752.1 | 5 | 551 | |||
XM_017018265.1 | XP_016873754.1 | 6 | 666 | |||
XM_017018266.1 | XP_016873755.1 | 5 | 586 | |||
XM_017018267.1 | XP_016873756.1 | 6 | 870 | |||
GenBank mRNA | AB209664.1 | - | 1 | 7342 | ||
AF003540.1 | - | 4 | 396 | |||
AK290099.1 | - | 4 | 442 | |||
AK308024.1 | - | 4 | 478 | |||
AK308209.1 | - | 4 | 427 | |||
BC121029.1 | - | 4 | 389 | 40117445 | ||
DB294374.1 | - | 6 | 586 |
