siRNA Details
| Gene Symbol: Nkg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133540.1 | NP_598224.1 | 4 | 676 | ||
| GenBank mRNA | AF082535.1 | - | 4 | 676 | ||
| FQ222255.1 | - | 4 | 682 | |||
| FQ226323.1 | - | 4 | 682 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkg7| Gene Symbol: Nkg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133540.1 | NP_598224.1 | 4 | 676 | ||
| GenBank mRNA | AF082535.1 | - | 4 | 676 | ||
| FQ222255.1 | - | 4 | 682 | |||
| FQ226323.1 | - | 4 | 682 | |||